missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM101B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556579
This item is not returnable.
View return policy
Description
Immunogen sequence: PNGTWRNYKA EVRFEPRHRP TRFLSTTIVY PKYPKAVYTT TLDYNCRKTL RRFLSSVELE AAELPGSDDL SDEC Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
RFLNB is a protein coding gene involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in the formation of cartilaginous skeletal elements.
Specifications
| FAM101B | |
| Polyclonal | |
| Unconjugated | |
| RFLNB | |
| 1500005K14Rik; cfm; Cfm1; Fam101b; family with sequence similarity 101 member B; family with sequence similarity 101, member B; filamin-interacting protein FAM101B; filamin-interacting protein FAM101B; refilin-B; MGC45871; protein FAM101B; refilin B; RefilinB; refilin-B; regulator of filamin protein B; RFLNB; RGD1359691 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 359845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N5W9 | |
| RFLNB | |
| Recombinant protein corresponding to Human FAM101B. Recombinant protein control fragment (Product #RP-93503). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction