missing translation for 'onlineSavingsMsg'
Learn More

FAM101B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-62633

 View more versions of this product

Product Code. 18616068

  • £513.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FAM101B Polyclonal antibody specifically detects FAM101B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAM101B
Polyclonal
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
CFM1, FAM101B, Family With Sequence Similarity 101, Member B, Filamin-Interacting Protein FAM101B, Protein FAM101B, Refilin B, RefilinB, Refilin-B, Regulator Of Filamin Protein B, RFLNB
This antibody was developed against a recombinant protein corresponding to amino acids: EGVEFDPLPPKEVRYTSLVKYDSERHFIDDVQLPLGLAVASCSQTVTCVP
100 μg
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Protein A purified
RUO
359845
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.