missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM101A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585048
This item is not returnable.
View return policy
Description
Immunogen sequence: MLPVFFGESI KVNPEPTHEI RCNSEVKYAS EKHFQDKVFY APVPTVTAYS ETIVAAPNCT WRNY Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 84%.
Involved in the regulation of the perinuclear actin network and nuclear shape through interaction with filamins. Plays an essential role in actin cytoskeleton formation in developing cartilaginous cells.
Specifications
| FAM101A | |
| Polyclonal | |
| Unconjugated | |
| RFLNA | |
| 3110032G18Rik; cfm; CFM2; FAM101A; family with sequence similarity 101 member A; family with sequence similarity 101, member A; filamin-interacting protein FAM101A; protein FAM101A; refilin A; RefilinA; refilin-A; regulator of filamin protein A; RFLNA; si:bz71m17.1; si:rp71-71m17.1; ZNF664-FAM101A; ZNF664-FAM101A readthrough | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 144347 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZTI6 | |
| RFLNA | |
| Recombinant protein corresponding to Human RFLNA. Recombinant protein control fragment (Product #RP-108296). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction