missing translation for 'onlineSavingsMsg'
Learn More

Faltan, MedChemExpress

Product Code. 16482359
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482359 Supplier Medchem Express Supplier No. HYB1878
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Faltan is a dicarboximide fungicide, widely used on vines and several vegetable crops, and is also cytotoxic effect on human bronchial epithelial cells.

Faltan is a dicarboximide fungicide, widely used on vines and several vegetable crops, and is also cytotoxic effect on human bronchial epithelial cells.

Specifications

Chemical Name or Material Faltan
Physical Form Solid
CAS 133-07-3
Color Light Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H4Cl3NO2S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (421.50 mM; Need ultrasonic)
SMILES O=C1N(SC(Cl)(Cl)Cl)C(C2=C1C=CC=C2)=O
Molecular Weight (g/mol) 296.56
Formula Weight 296.56
Percent Purity 98.55%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.