missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific False Bottom Tubes for Automation

Product Code. 31923236 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Natural
White
Product Type:
False Bottom Tube
Packaging:
1000/Cs.
1500/Cs.
Unit Size:
Pack of 1000
Pack of 1500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Packaging Unit Size
31923236 White False Bottom Tube 1500/Cs. Pack of 1500
31922516 Natural False Bottom Tube 1000/Cs. Pack of 1000
31922497 Natural False Bottom Tube 1500/Cs. Pack of 1500
31922679 White False Bottom Tube 1000/Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31923236 Supplier Globe Scientific Supplier No. 5528B
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as secondary tube for barcoding on automated analyzers

 

  • Made of Polypropylene
  • Dimensions: 16 x 97mm
  • Capacity: 5mL
  • Screw Caps are sold separately (See 21-200-231, -232)

 

Specifications

Color White
Description These are the screw caps for use with 5527 and 5527B False Bottom Tubes
Material Polypropylene
Packaging 1500/Cs.
Product Type False Bottom Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.