missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Standard Tissue Culture Dishes

Product Code. 11527411 Shop All Corning Products
Change view
Click to view available options
Culture Area:
21.29 cm2
58.1 cm2
Volume (Metric) Working:
16 to 17.5 mL
6 to 7 mL
Unit Size:
Pack of 200
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Volume (Metric) Working unitSize
11527411 21.29 cm2 6 to 7 mL Pack of 500
10212951 58.1 cm2 16 to 17.5 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11527411 Supplier Falcon™ Supplier No. 353002
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.

  • Flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells
  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene
  • Standard Tissue Culture (TC), Corning™ PrimariaTM surface treatment and non-TC-treated polystyrene available

Designed for cell culture

  • Flat, distortion-free optics
  • Lids designed for optimal gas exchange
  • Stacking rings allow for easier stacking and handling
  • Vacuum-gas plasma treatment permanently and consistently modifies the cell growth surface
  • Standard tissue culture surface is hydrophilic and contains a variety of negatively charged functional groups that support cell attachment and spreading
  • Corning Primaria tissue culture surface additionally incorporates nitrogen-containing functional groups and has been shown to support improved attachment, spreading, and differentiation of some cell types

Order Info

Packed 20 per sleeve.

TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Flat
Capacity (Metric) 7 mL (Well)
Color Clear
Culture Area 21.29 cm2
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Cell Culture
Type Cell Culture Petri Dish
Volume (Metric) Working 6 to 7 mL
No. per Case 500/Cs.
No. per Pack 20/Pk.
Height (Metric) 15 mm
Diameter (Metric) 60 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.