missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ platelet-derived growth factor-BB (PDGF-BB), human recombinant, 100μg

Product Code. 10584461 Shop All Corning Products
Change view
Click to view available options
Quantity:
10 x 10 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10584461 10 x 10 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10584461 Supplier Corning Supplier No. 356051

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Corning Platelet-Derived Growth Factor-BB (PDGF-BB), human recombinant, is a dimeric glycoprotein composed of two A and/or B chains. This growth factor is the principal mitogen in serum for mesenchymal cells.
  • Formulation: lyophilised from 10 mM acetic acid, carrier-free; reconstitute in 10 mM acetic acid or serum-free medium containing 0.2% BSA
  • Sourced from recombinant E. coli
  • Molecular weight 25kD
  • Purity more than 95% by SDS-PAGE
  • Tested for ability to stimulate proliferation of Swiss albino mouse fibroblasts
  • Filtered (0.2μm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Recommended concentration 0.5 to 4 ng/mL of medium depending on cell type
  • Stable for at least three months at 2° to 8°C (lyophilised) or three months at -20°C (solubilised). Avoid multiple freeze/thaws
TRUSTED_SUSTAINABILITY

Specifications

Purity or Quality Grade >95% by SDS-PAGE
Quantity 10 x 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.