missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ HTS 24-Well Multiwell Permeable Support System
GREENER_CHOICE

Product Code. 10326011 Shop All Corning Products
Change view
Click to view available options
Pore Size:
1 μm
3 μm
8 μm
Quantity:
5/Cs.
Each
Unit Size:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Quantity unitSize
10326011 3 μm 5/Cs. Pack of 5
10182511 8 μm 5/Cs. Pack of 5
10693462 8 μm Each Each
10265141 1 μm 5/Cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10326011 Supplier Falcon™ Supplier No. 351183

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

24-well insert plate, 24-well plate or single-well feeder tray, and lid

Save time and labor in HTS protocols—grow, handle, and analyze 24 samples simultaneously. Falcon™ HTS Multiwell Insert System features a generous, automation-compatible sampling port which allows sampling above and below membranes.

Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Color Clear
Culture Area 0.3 cm2
Material PET
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Insert System
Well Count 24
For Use With (Application) Excellent for differentiation, toxicology, migration and invasion studies
Includes 24-well insert plate, 24-well plate and lid
No. per Case 5/Cs.
No. per Pack 5/Pk.
Pore Size 3 μm
Quantity 5/Cs.
Supportive Membrane Polyethylene Terephthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.