Learn More
Description
Includes
24-well insert plate, 24-well plate or single-well feeder tray, and lid
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
Specifications
Specifications
| Closure Included | Yes |
| Color | Clear |
| Culture Area | 0.3 cm2 |
| Material | PET |
| Sterility | Sterile |
| Surface Treatment | Cell Culture |
| Type | Multi-well Insert System |
| Well Count | 24 |
| For Use With (Application) | Excellent for differentiation, toxicology, migration and invasion studies |
| Includes | 24-well insert plate, 24-well plate and lid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
