missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Companion Plates for Cell Culture Inserts
GREENER_CHOICEBIOPHARMA

Product Code. 10386881 Shop All Corning Products
Change view
Click to view available options
Culture Area:
3.8 cm2
9.6 cm2
Volume (Metric) Working:
1.4 to 2.3 mL
2.7 to 3.2 mL
Well Count:
6
12
Capacity (Metric):
2.3 mL (Well)
3.2 mL (Well)
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Volume (Metric) Working Well Count Capacity (Metric) unitSize
10386881 9.6 cm2 2.7 to 3.2 mL 6 3.2 mL (Well) Pack of 50
10389270 3.8 cm2 1.4 to 2.3 mL 12 2.3 mL (Well) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10386881 Supplier Falcon™ Supplier No. 353502
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Feature a one-way, labyrinth lid design and condensation rings to minimize evaporation and contamination. Falcon™ Companion Plates for Cell Culture Inserts are specifically designed for use with Falcon or Corning™ BioCoat™ Cell Culture Inserts.

Falcon Cell Culture Insert Companion Plates have been specially designed for use with Falcon or Corning™ BioCoatTM Cell Culture Inserts so evaporation and contamination due to improper lid fit is eliminated.

In the Feeding Position, pipet access is improved for fluid handling on the basolateral side. Reagents can be added quickly and consistently for timed experiments. Aspiration of media from the well is easier, reducing the risk of contamination.

In the Incubation Position, Falcon Cell Culture Inserts remain locked in position in their Falcon Companion Plate wells. Media will not wick up between the insert and well wall. The unique Falcon low-evaporation lid provides a tortuous air passage system that reduces evaporation and contamination.

TRUSTED_SUSTAINABILITY

Specifications

Closure Included Yes
Capacity (Metric) 3.2 mL (Well)
Color Clear
Culture Area 9.6 cm2
Material Polystyrene
Sterility Sterile
Surface Treatment Cell Culture
Type Multi-well Insert System
Volume (Metric) Working 2.7 to 3.2 mL
Well Count 6
For Use With (Equipment) Cell culture inserts
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.