missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Cell Culture Inserts: For Use With 6-Well Plates
GREENER_CHOICEBIOPHARMA

Product Code. 10356981 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
Type:
Translucent Membrane
Transparent Membrane
Unit Size:
Pack of 48
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Type unitSize
10356981 0.4 μm Translucent Membrane Pack of 48
10380291 0.4 μm Transparent Membrane Pack of 48
10768882 8 μm Translucent Membrane Pack of 48
10758882 3 μm Transparent Membrane Pack of 48
10025090 3 μm Translucent Membrane Pack of 48
10289270 1 μm Transparent Membrane Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10356981 Supplier Falcon™ Supplier No. 353493

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cell culture on permeable supports results in greater morphological and/or physiological differentiation of many cell types

  • Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12, and 24 well
  • A broad selection of membrane pore sizes, 0.4, 1.0, 3.0, and 8.0 μm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture

Order Info

Individually packaged in color-coded, peel-open medical-style packaging

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 4.2 cm2
Material PET
Pore Density 1.0 x 108 (HD)
Pore Size 0.4 μm
Type Translucent Membrane
For Use With (Equipment) 6 Well Plate
Diameter (Metric) Filter 23.1 mm
No. per Pack 6/Pk.
No. per Case 48/Cs.
Surface Coating TC-Treated
Surface Treatment TC-Treated
Sterility Sterile
No. of Wells 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.