missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ FluoroBlok™ HTS 96-Well Insert Systems
GREENER_CHOICE

Codice prodotto. 10316182 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Pore Size:
3 μm
8 μm
Quantity:
1/Cs.
5/Cs.
Dimensione della confezione:
Each
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Pore Size Quantity unitSize
10316182 3 μm 1/Cs. Each
10172822 3 μm 5/Cs. Pack of 5
10520613 8 μm 1/Cs. Each
10530613 8 μm 5/Cs. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10316182 Fornitore Falcon™ N. del fornitore 351161
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Simplify detection of migrating fluorescently labeled cells. Falcon™ FluoroBlok™ 96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.

  • A solution for automated, high throughput cell-based studies of chemotaxis, migration and invasion
  • Increase cell migration and invasion assay productivity with simplified fluorescence detection and real-time analysis
  • Simplification of chemotaxis, cell migration, and invasion assays
  • Standard technology platform allows multiple protocols
  • Homogeneous protocols for real-time kinetic readouts
  • Real-time detection without dismantling or destroying the insert
  • Increased sample throughput
  • Eliminates need for manual cell scraping and counting
  • Automation friendly
  • 96 Multiwell format is compatible with commercial detectors and fluid handling instruments
  • Unique fluorescence blocking membrane
  • Blocks greater than 99% of the excitation and emission wavelengths of fluorophores commonly used to label cells

The Corning FluoroBlok 96 Multiwell Insert System is a cell culture assay platform designed with automation in mind. The one-piece insert housing and fluorescence blocking microporous membrane (available in 3.0 and 8.0 μm pore sizes) enables increased efficiency, productivity and throughput in the drug discovery process. The novel receiver plate design minimizes crosstalk between the wells; the black housing of the 96 Multiwell Insert virtually eliminates autofluorescence. These features ensure fluorescence measurements that result from your assay, not crosstalk or background signal.

Order Info

Available in 3, and 8μm pore sizes. One-piece 96-Multiwell insert system (insert plate, 24-well plate or single-well feeder tray, and lid) or 24-well individual cell culture inserts.

TRUSTED_SUSTAINABILITY

Specifica

Format Standard
Lid With Lid
Culture Area 0.08 cm2
Membrane PET
Array 8 x 12
Pore Size 3 μm
Color Black
Product Type FluoroBlok™ HTS 96-Well Multiwell Permeable Support System
For Use With (Equipment) Fluorescence plate reader or inverted fluorescence microscope
Material PET
No. of Wells 96
Sterility Sterile
Includes Insert plate with 96 square well, flat bottom plate and lid
No. per Case 1/Cs.
No. per Pack 1/Pk.
Quantity 1/Cs.
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.