missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Feeder Tray for FluoroBlok™ HTS 24 Well Multiwell plates
GREENER_CHOICE

Product Code. 10192891 Shop All Corning Products
Change view
Click to view available options
Well Count:
24
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count unitSize
10192891 24 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192891 Supplier Falcon™ Supplier No. 351186
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polystyrene cell media tray for feeding cell-based assays.

Polystyrene cell media tray for feeding cell-based assays

Storage: Store at room temperature, away from direct sunlight.

TRUSTED_SUSTAINABILITY

Specifications

Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Non-treated
Type Feeder Tray
Well Count 24
For Use With (Equipment) FluoroBlok™ HTS 24 Well Multiwell plates, Falcon™ 24 Well Multiwell plates
Includes Lid
No. per Case 5/Cs.
No. per Pack 5/Pk.
Pore Size Not applicable
Quantity 5/Pk.
Storage Requirements Store at room temperature, away from direct sunlight
Supportive Membrane Polyethylene Terephthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.