missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX FAK Protein, Active

Product Code. 17390604
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17390604

Brand: Eurofins DiscoverX 14720M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal 6His-tagged, recombinant, human FAK, amino acids 411-686, for use in Kinase Assays.

PTK2 (Protein Tyrosine Kinase 2, formerly FAK or Focal Adhesion Associated Protein Tyrosine Kinase) is a widely expressed nonreceptor involved in the structure and function of focal adhesions, which are critical for cellular integrity by maintaining cell-cell and cell-matrix interactions. Integrin-mediated cell adhesion causes activation of MAP kinases and increased tyrosine phosphorylation of PTK2. Autophosphorylation of PTK2 leads to the binding of SH2-domain proteins including Src-family kinases and the Grb2-Sos complex, then activate Ras cascade and ultimately to mitogen activated protein (MAP) kinase activation.

GenBank L13616

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 35.5 kDa
Quantity 250 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥68%
Protein FAK, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.