missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAIM2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | FAIM2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348744
|
Novus Biologicals
NBP3-04403-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18335414
|
Novus Biologicals
NBP3-04403-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FAIM2 Polyclonal specifically detects FAIM2 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FAIM2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Fas apoptotic inhibitory molecule 2, KIAA0950NGP35, LFG2, LFGLIFEGUARD, NMP35neural membrane protein 35, Protein lifeguard, TMBIM2lifeguard, transmembrane BAX inhibitor motif containing 2, Transmembrane BAX inhibitor motif-containing protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human FAIM2 (NP_036438.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS with 50% glycerol, pH7.3. | |
| 23017 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title