missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAHD2B Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | FAHD2B |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FAHD2B Polyclonal specifically detects FAHD2B in Mouse samples. It is validated for Western Blot.Specifications
| FAHD2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| DKFZp434N062, EC 3.-, fumarylacetoacetate hydrolase domain containing 2B, fumarylacetoacetate hydrolase domain-containing protein 2B | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Mouse FAHD2B (NP_083905). Peptide sequence VSQFVTLYPGDLLLTGTPPGVGMFRKPPVFLKKGDEVQCEIEELGVIINK | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 151313 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title