missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody (OTI10D1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71679B
This item is not returnable.
View return policy
Description
FAF1 Monoclonal specifically detects FAF1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FAF1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| FAF1 | |
| Recombinant protein fragment corresponding to amino acids 415-650 of human FAF1 (NP_008982) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI10D1 | |
| Western Blot | |
| CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 11124.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction