missing translation for 'onlineSavingsMsg'
Learn More

FAF1 Antibody (CPTC-FAF1-2) - Azide and BSA Free, Novus Biologicals™

Product Code. 18395393 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395393 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395393 Supplier Novus Biologicals Supplier No. NBP308227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FAF1 Monoclonal specifically detects FAF1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAF1
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone CPTC-FAF1-2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1
Host Species Mouse
Immunogen Recombinant human full-length FAF1 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11124
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.