missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | FAF1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
FAF1 Polyclonal specifically detects FAF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FAF1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer | |
| PBS with 0.02% Sodium Azide | |
| CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
| FAF1 | |
| IgG | |
| Affinity Purified | |
| 80 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_008982 | |
| 11124 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FAF1. The immunogen is located within the last 50 amino acids of FAF1. Amino Acid Squence: STFPRRDVTQLDPNKSLLE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title