missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF1 Antibody (9H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42969
This item is not returnable.
View return policy
Description
FAF1 Monoclonal antibody specifically detects FAF1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| FAF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| 9H10 | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 | |
| Fas(TNFRSF6) associated factor 1 - (Uniprot# Q9UNN5) | |
| 100 μg | |
| Apoptosis, Cancer | |
| 11124 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction