missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FADS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548353
This item is not returnable.
View return policy
Description
Predicted to react with monkey based on sequence homology.
The protein encoded by this gene is a member of the fatty acid desaturase (FADS) gene family. Desaturase enzymes regulate unsaturation of fatty acids through the introduction of double bonds between defined carbons of the fatty acyl chain. FADS family members are considered fusion products composed of an N-terminal cytochrome b5-like domain and a C-terminal multiple membrane-spanning desaturase portion, both of which are characterized by conserved histidine motifs. This gene is clustered with family members FADS1 and FADS2 at 11q12-q13. 1; this cluster is thought to have arisen evolutionarily from gene duplication based on its similar exon/intron organization.
Specifications
| FADS2 | |
| Polyclonal | |
| Unconjugated | |
| Fads2 | |
| 2900042M13Rik; Acyl-CoA 6-desaturase; D6D; delta(6) desaturase; delta(6) fatty acid desaturase; delta-6 desaturase; delta-6 fatty acid desaturase; delta-6-desaturase; DES6; FADS2; Fadsd2; FADSD6; Fatty acid desaturase 2; linoleoyl-CoA desaturase (delta-6-desaturase)-like 2; LLCDL2; SLL0262; TU13 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 9415 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.48 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O95864 | |
| Fads2 | |
| KLH conjugated synthetic peptide between 79-108 amino acids from the N-terminal region of human FADS2. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction