missing translation for 'onlineSavingsMsg'
Learn More

Fadrozole, MedChemExpress

Product Code. 16496416
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496416 100 mg 100mg
16488617 5 mg 5mg
16433416 25 mg 25mg
16422147 50 mg 50mg
16406008 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496416 Supplier Medchem Express Supplier No. HY14247A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.

Fadrozole (CGS 16949A free base) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.

Specifications

Chemical Name or Material Fadrozole
CAS 102676-47-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C14H13N3
Quantity 100 mg
Synonym CGS 16949A free base (Rac)-FAD286
Solubility Information DMSO : ≥ 100 mg/mL (447.89 mM)
SMILES N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1
Molecular Weight (g/mol) 223.27
Formula Weight 223.27
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.