missing translation for 'onlineSavingsMsg'
Learn More

Fadrozole Hydrochloride, TRC

Product Code. 30393089
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393089 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393089 Supplier Toronto Research Chemicals Supplier No. TRCF10104825MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fadrozole Hydrochloride
CAS 102676-31-3
Molecular Formula C14H13N3 . HCl
Quantity 25 mg
Synonym Imidazo[1,5-a]pyridine, benzonitrile deriv., Afema, CGS 16949, CGS 16949A, 4-(5,6,7,8-tetrahydroimidazo[1,5-a]pyridin-5-yl)benzonitrile
InChI Formula InChI=1S/C14H13N3.ClH/c15-8-11-4-6-12(7-5-11)14-3-1-2-13-9-16-10-17(13)14;/h4-7,9-10,14H,1-3H2;1H
SMILES Cl.N#Cc1ccc(cc1)C2CCCc3cncn23
Molecular Weight (g/mol) 259.73
Formula Weight 259.0876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.