missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fadrozole hydrochloride, MedChemExpress
Description
Fadrozole hydrochloride (CGS 16949A) is a potent, selective and nonsteroidal inhibitor of aromatase with an IC50 of 6.4 nM.
Specifications
Specifications
| Chemical Name or Material | Fadrozole hydrochloride |
| CAS | 102676-31-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H14ClN3 |
| Quantity | 10 mg |
| Synonym | CGS 16949A(Rac)-FAD286 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (385.02 mM; Need ultrasonic and warming) ∣H2O : 100 mg/mL (385.02 mM; Need ultrasonic) |
| SMILES | N#CC1=CC=C(C2CCCC3=CN=CN23)C=C1.Cl |
| Molecular Weight (g/mol) | 259.73 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?