missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£189.00 - £380.00
Specifications
| Antigen | FADD |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383694
|
Novus Biologicals
NBP3-03350-20UL |
20 μL |
£189.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18323586
|
Novus Biologicals
NBP3-03350-100UL |
100 μL |
£380.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FADD Polyclonal specifically detects FADD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| FADD | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human FADD (NP_003815.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors | |
| PBS with 50% glycerol, pH7.3. | |
| 8772 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title