missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Monoclonal Antibody (OTI1C11)

FADD Monoclonal Antibody (OTI1C11)
Brand: Invitrogen MA525179
This item is not returnable.
View return policy
Description
Specifications
| FADD | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13158 | |
| FADD | |
| Full length human recombinant protein of FADD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI1C11 | |
| Unconjugated | |
| FADD | |
| Fadd; Fas (TNFRSF6)-associated via death domain; Fas associated via death domain; FAS-associated death domain protein; FAS-associating death domain-containing protein; Fas-associating protein with death domain; GIG3; Growth-inhibiting gene 3 protein; mediator of receptor induced toxicity; mediator of receptor-induced toxicity; MGC8528; MORT1; Mort1/FADD; protein FADD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8772 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction