missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Antibody (PSH01-13), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32329
This item is not returnable.
View return policy
Description
FADD Monoclonal antibody specifically detects FADD in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| FADD | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
| Recombinant protein within human FADD aa 1-208/ 208. (Uniprot: Q13158) | |
| 100 μL | |
| Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors | |
| 8772 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PSH01-13 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction