missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FADD Antibody (OTI1C11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71387AF532
This item is not returnable.
View return policy
Description
FADD Monoclonal specifically detects FADD in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| FADD | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FADD | |
| Full length recombinant protein of human FADD (NP_003815) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI1C11 | |
| Western Blot, Flow Cytometry | |
| Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
| Mouse | |
| 0.1 mL | |
| 8772 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction