missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FADD Monoclonal antibody specifically detects FADD in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | FADD |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1A11 |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD |
| Host Species | Mouse |
| Immunogen | The immunogen is a peptide made to the amino acid region FADD |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?