missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Mouse anti-Human, Clone: F13A1/1448, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59604-20UG
This item is not returnable.
View return policy
Description
Factor XIIIa Monoclonal specifically detects Factor XIIIa in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Factor XIIIa | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5-1.0 ug/ml, Flow Cytometry 0.5-1 ug/million cells, ELISA 2-4 ug/ml, Immunocytochemistry/Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page, Protein Array 1:100-1:2000 | |
| bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
| Mouse | |
| 83 kDa | |
| 20 μg | |
| Apoptosis, Cancer, Cell Biology | |
| 2162 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| F13A1/1448 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| F13A1 | |
| Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. It recognizes a protein of 83kDa, which is identified as Factor XIIIa. It has been identified in platelets, megakaryocytes, and fibroblast-like mesenchymal or histiocytic cells in the placenta, uterus, and prostate, monocytes and macrophages and dermal dendritic cells. Anti-factor XIIIa has been found to be useful in differentiating between dermatofibroma (almost all cases are positive), dermatofibrosarcoma protuberans ( -/+) and desmoplastic malignant melanoma (-). Anti-factor XIIIa positivity is also seen in capillary hemagioblastoma, hemangioendothelioma, hemangiopericytoma, xanthogranuloma, xanthoma, hepatocellular carcinoma, glomus tumor, and meningioma. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction