missing translation for 'onlineSavingsMsg'
Learn More

Factor XIIIa Antibody (F13A1/1448), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18431105 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431105 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431105 Supplier Novus Biologicals Supplier No. NBP259605MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Factor XIIIa Monoclonal antibody specifically detects Factor XIIIa in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XIIIa
Applications Flow Cytometry
Classification Monoclonal
Clone F13A1/1448
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma
Host Species Mouse
Immunogen Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) (Uniprot: P00488)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 2162
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.