missing translation for 'onlineSavingsMsg'
Learn More

Factor XII Antibody (OTI5A6) [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30688006 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30688006 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30688006 Supplier Novus Biologicals Supplier No. NBP344901AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Factor XII Monoclonal antibody specifically detects Factor XII in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XII
Applications Western Blot
Classification Monoclonal
Clone OTI5A6
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 73-372 of human F12(NP_000496) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2161
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.