missing translation for 'onlineSavingsMsg'
Learn More

Factor XII Antibody (10-11-37), Novus Biologicals™

Product Code. 18356913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356913 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356913 Supplier Novus Biologicals Supplier No. NBP311299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Factor XII Monoclonal specifically detects Factor XII in Human, Mouse samples. It is validated for Western Blot, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor XII
Applications Western Blot, Immunoassay
Classification Monoclonal
Clone 1937-10-11 00:00:00
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, F12, HAE3, HAEX, HAF, HAF beta-factor XIIa part 2, Hageman factor
Host Species Mouse
Immunogen Factor XII adsorbed onto Al(OH)3 adjuvant and emulsified in Freund's Incomplete Adjuvant
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2161
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.