missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor X/Xa Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA143061
This item is not returnable.
View return policy
Description
PA1-43061 detects Factor X in mouse samples. PA1-43061 has successfully been used in Western blot and ELISA applications. PA1-43061 was derived from mouse factor X.
Factor X undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
Specifications
| Factor X/Xa | |
| Polyclonal | |
| Unconjugated | |
| F10 | |
| Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product | |
| Sheep | |
| SDS-PAGE | |
| RUO | |
| 14058, 29243 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 25 mg/mL | |
| 50% water with 50% glycerol and no preservative | |
| O88947, Q63207 | |
| F10 | |
| Murine Factor X. | |
| 1 mg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction