Learn More
Invitrogen™ Factor X/Xa Monoclonal Antibody (5050)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA143013
Description
This antibody detects the heavy chains of human factor Xa and X; does not bind bovine factor X or BEGRck-Xa. Does not recognize murine or porcine X via Western blot. Inhibitory properties: inhibits PT, prothrombinase and partially APTT, but not amidase activity. Purity >95% by SDS-PAGE.
Factor X undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
Specifications
| Factor X/Xa | |
| Monoclonal | |
| 7.6 mg/mL | |
| 50% water with 50% glycerol and no preservative | |
| P00742 | |
| F10 | |
| Purified human Factor X. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Affinity Purification, Inhibition Assays, Inhibition Assays, Radioimmune Assays (RIA), Western Blot | |
| 5050 | |
| Unconjugated | |
| F10 | |
| Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product | |
| Mouse | |
| SDS-PAGE | |
| RUO | |
| 2159 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.