missing translation for 'onlineSavingsMsg'
Learn More

Factor VIII Antibody (610), DyLight 488, Novus Biologicals™

Product Code. 18352490 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352490 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352490 Supplier Novus Biologicals Supplier No. NBP290256G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Factor VIII Monoclonal specifically detects Factor VIII in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor VIII
Applications ELISA
Classification Monoclonal
Clone 610
Conjugate DyLight 488
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Factor VIII.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 2157
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.