missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor VII Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547475
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Factor VII, rhFactor X, and rhFactor Xa is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
Specifications
| Factor VII | |
| Polyclonal | |
| Unconjugated | |
| F7 | |
| AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 14068 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P70375 | |
| F7 | |
| CHO-derived recombinant mouse Coagulation Factor VII Ala42-Leu446. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction