Learn More
Invitrogen™ Factor VII Monoclonal Antibody (321621)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561141
Product Code. 30536142
  • £357.00 / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Factor VII
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P08709
F7
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII, Ala39-Pro444.
100 μg
Primary
Human
Antibody
IgG1
Immunoprecipitation, Western Blot, Immunocytochemistry
321621
Unconjugated
F7
AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA
Mouse
Protein A/G
RUO
2155
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized