missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor V Monoclonal Antibody (5104)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA143007
This item is not returnable.
View return policy
Description
MA1-43007 detects Factor V in bovine samples. MA1-43007 has successfully been used in RIA, WB, ELISA, IA applications. MA1-43007 was derived from bovine factor Va heavy chain.
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
Specifications
| Factor V | |
| Monoclonal | |
| 9.5 mg/mL | |
| 50% water with 50% glycerol and no preservative | |
| Q28107 | |
| F5 | |
| Bovine factor Va heavy chain. | |
| 100 μg | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG1 |
| ELISA, Inhibition Assays, Radioimmune Assays (RIA), Western Blot | |
| 5104 | |
| Unconjugated | |
| F5 | |
| Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2 | |
| Mouse | |
| SDS-PAGE | |
| RUO | |
| 280687 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction