missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress α-Factor Mating Pheromone, yeast

Product Code. 16492919
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492919 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492919 Supplier MedChemExpress Supplier No. HYP1482
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress α-Factor Mating Pheromone, yeast is a tridecapeptide secreted by S. cerevisiae α cells via Ste2p receptor.

Specifications

Purity 99.8%
CAS 59401-28-4
Content And Storage Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
For Use With (Application) Metabolism-protein/nucleotide metabolism
Molecular Weight (g/mol) 1684.0 g/mol
Product Type Natural Products
Quantity 1 mg
Shipping Condition Room temperature
Solubility H2O : 50 mg/mL (29.69 mM∣Need ultrasonic)
Synonym Mating Factor α
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.