Learn More
Invitrogen™ Factor IX Recombinant Rabbit Monoclonal Antibody (044)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA530583
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Coagulation Factor IX/FIX/F9.
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifications
| Factor IX | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P00740 | |
| F9 | |
| Recombinant Human Coagulation Factor IX/FIX/F9 Protein (Met1-Thr461). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 044 | |
| Unconjugated | |
| F9 | |
| AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.