missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Factor IX Monoclonal Antibody (C23), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA551102

 View more versions of this product

Product Code. 17939912

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 μg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Factor IX
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P00740
F9
Recombinant Human F9 protein, Asp232-Lys455 (Accession #P00740).
20 μL
Primary
Human
Antibody
IgG2a κ
Western Blot, Immunohistochemistry (Paraffin), Western Blot
C23
Unconjugated
F9
AW111646; Cf9; Cf-9; Christmas factor; coagulation factor 9; coagulation factor IX; coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); Coagulation factor IXa heavy chain; Coagulation factor IXa light chain; EC 3.4.21.22; F9; F9 p22; factor 9; factor IX (Christmas factor) precursor; factor IX F9; Factor9; FIX; FIX F9; HEMB; P19; plasma thromboplastic component; plasma thromboplastin component; PTC; RP6-88D7.1; serine protease; THPH8
Mouse
Protein A/G
RUO
2158
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.