missing translation for 'onlineSavingsMsg'
Learn More

Factor IX Antibody (034), DyLight 488, Novus Biologicals™

Product Code. 18341540 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341540 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341540 Supplier Novus Biologicals Supplier No. NBP289906G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Factor IX Monoclonal specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Factor IX
Applications Western Blot, ELISA
Classification Monoclonal
Clone 34
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Factor IX (AAB59620.1; Met 1-Thr 461).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2158
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.