missing translation for 'onlineSavingsMsg'
Learn More

Ekastu Safety™ Mandil Filtering Facepiece FFP1

Product Code. 10320411
Click to view available options
Description:
Filter class DIN EN 149: 2001 FFP1 NR
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 10320411

Brand: Ekastu Safety™ 411110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Offers protection against fine inert particles up to 4.5 times the NPF value. EKASTU Safety™ Mandil Filtering Facepiece FFP1 has a five-year shelf life in original packaging.

  • With color-coded nosepiece
  • Foam lined comfort nose zone
  • Protection against fine inert particles up to 4.5 times the NPF value
  • Filter class DIN EN 149:2001 FFP1 NR
  • Specially designed for wearer of EKASTU spectacles and goggles
  • In original packaging: 5 years shelf life
  • 12 pieces per pack

Specifications

Color White
Size Standard
Certifications/Compliance DIN EN 149:2001 FFP1 NR
Description Filter class DIN EN 149: 2001 FFP1 NR
Disposable Single-use
For Use With (Application) Protection against fine inert particles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.