Learn More
Ansell Edmont™ Fabric Brown Over Sleeves
Product Code. 12980645 Shop All Ansell Products
Change view
Click to view available options
Length (Metric):
66 cm
Product Code. Length (Metric)
12980645 66 cm
1 options
Product Code. 12980645 Supplier Ansell Edmont™ Supplier No. 59416
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Welder sleeve made of Kevlar™ fabric – combined with an internal cotton layer. These make the sleeve lightweight and breathable while providing protection from the high heat of flying molten particles and sparks.
  • Heat and cut resistant protective sleeve for welders
  • Protection from high heat and flying molten particles and sparks
  • Lightweight and breathable due to the internal cotton layer
TRUSTED_SUSTAINABILITY
Specifikationer
Size Standard
Construction Cut and sewn
Product Type Welder Sleeve
Material Dupont™ Kevlar™ aramid fibers
Color Brown
Sterility Non-sterile
Style Sleeve with Thumb Hole
Disposable Reusable
Closure Type Elastic Upper with Knit Cuff
Latex Free No
Certifications/Compliance EN 388, 407, CE 0493
Length (English) 26 in. (66 cm)
Length (Metric) 66 cm
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.