missing translation for 'onlineSavingsMsg'
Learn More

FABP8/M-FABP/Myelin P2 Protein Antibody (001), Janelia Fluor™ 646 , Novus Biologicals™

Product Code. 18405919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405919 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405919 Supplier Novus Biologicals Supplier No. NBP289897JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP8/M-FABP/Myelin P2 Protein
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP8/M-FABP/Myelin P2 Protein (NP_002668.1; Ser 2-Val 132).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5375
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.