missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP8/M-FABP/Myelin P2 Protein Antibody (001), Alexa Fluor™ 594 , Novus Biologicals™
Shop All Bio Techne ProductsDescription
FABP8/M-FABP/Myelin P2 Protein Monoclonal antibody specifically detects FABP8/M-FABP/Myelin P2 Protein in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | FABP8/M-FABP/Myelin P2 Protein |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 001 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | M-FABP, MP2, myelin P2 protein, P2, peripheral myelin protein 2FABP8 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human FABP8/M-FABP/Myelin P2 Protein (NP_002668.1; Ser 2-Val 132). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?