missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FABP7 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA524949

Product Code. 13292749

  • £485.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with bovine and chicken based on sequence homology.

FABP7 was initially isolated from a human fetal brain cDNA library and whose mRNA was expressed in adult brain and muscle tissues at low levels. The protein encoded by this gene is a member of the fatty acid binding protein (FABPs) family, a group of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABPs are thought to play roles in fatty acid uptake, transport, and metabolism. FABP7 is a downstream gene of the Pax6 transcription factor and has been suggested to be essential for the maintenance of neuroepithelial cells during early cortical development. More recently, FABP7 was found to be frequently expressed in melanomas. Down-regulation of FABP7 through RNAi expression could reduce in vitro cell proliferation and Matrigel invasion, suggesting that FABP7 may be a potential target for the development of diagnostic and therapeutic tools.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FABP7
Polyclonal
Unconjugated
FABP7
B FABP; BFABP; B-FABP; BLBP; brain lipid binding protein; brain lipid-binding protein; Brain-type fatty acid-binding protein; DKFZp547J2313; FABP 7; FABP7; FABPB; fatty acid binding protein 7; fatty acid binding protein 7, brain; fatty acid-binding protein 7; Fatty acid-binding protein, brain; fatty acid-binding protein, brain-like protein; hypothetical protein DKFZp547J2313; Mammary-derived growth inhibitor related; mammary-derived growth inhibitor-related; MRG; OTTHUMP00000017119; RP11-548E19.1
Rabbit
Antigen affinity chromatography, Protein A
RUO
12140, 2173
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O15540, P51880
FABP7
KLH conjugated synthetic peptide between 104-132 amino acids from the C-terminal region of human FABP7.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.