missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP7/B-FABP Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
£279.00 - £401.00
Specifications
| Antigen | FABP7/B-FABP |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649528
|
Novus Biologicals
NBP3-05531-100ul |
100 μg |
£401.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690407
|
Novus Biologicals
NBP3-05531-50ul |
50 μg |
£279.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP7/B-FABP Monoclonal antibody specifically detects FABP7/B-FABP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FABP7/B-FABP | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related | |
| Recombinant full length human FABP7/B-FABP expressed in and purified from E. coli | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Stem Cells | |
| 50% PBS, 50% glycerol | |
| 2173 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title