missing translation for 'onlineSavingsMsg'
Learn More

FABP7/B-FABP Antibody, Novus Biologicals™

Product Code. 18375682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375682 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375682 Supplier Novus Biologicals Supplier No. NBP311276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FABP7/B-FABP Polyclonal specifically detects FABP7/B-FABP in Human samples. It is validated for Western Blot, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FABP7/B-FABP
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias B-FABPDKFZp547J2313, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related
Host Species Rabbit
Immunogen The polyclonal antibody recognizes human brain fatty acid binding protein (B-FABP) of both natural and recombinant origin.
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2173
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.